The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032133.4 | NP_115509.4 | 14 - 15 | 2459 | 73 |
XM_005257725.3 | XP_005257782.1 | 14 - 15 | 2347 | 73 | |
XM_005257726.2 | XP_005257783.1 | 14 - 15 | 2347 | 73 | |
XM_017025205.1 | XP_016880694.1 | 14 - 15 | 2347 | 73 | |
GenBank mRNA | AB083068.1 | - | 14 - 15 | 2252 | 73 |
AK127992.1 | - | 11 - 12 | 1726 | 73 | |
AK303041.1 | - | 14 - 15 | 2459 | 73 | |
AK314052.1 | - | 14 - 15 | 2234 | 73 | |
AL136765.1 | - | 10 - 11 | 1601 | 73 | |
AL832225.1 | - | 14 - 15 | 2221 | 73 | |
BC028393.2 | - | 14 - 15 | 2260 | 73 |