Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DGCR9
- Entrez Gene ID
-
25787
- Gene Name
- DiGeorge syndrome critical region gene 9 (non-protein coding)
- Gene Aliases
-
DGS-A
- Chromosome Location
-
Chr.22: 19017834 - 19020248 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024159.1 |
|
1 - 1 |
1229 |
58 |
GenBank mRNA |
BC036821.1 |
- |
1 - 1 |
1240 |
58 |
L77571.1 |
- |
1 - 1 |
1127 |
58 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
DiGeorge syndrome critical region gene 5 (non-protein coding) |
DGCR5 |
LINC00037, NCRNA00037 |
|
Sense |
Chr. - 18970498 - 19031242 on Build GRCh38 |
Downstream |
DiGeorge syndrome critical region gene 10 (non-protein coding) |
DGCR10 |
DGS-B |
2.4kb
|
Sense |
Chr. - 19022624 - 19023550 on Build GRCh38 |
View Full Product Details