The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320941.1 | NP_001307870.1 | 18 - 19 | 2561 | 114 |
NM_001320942.1 | NP_001307871.1 | 19 - 20 | 2885 | 114 | |
NM_004136.3 | NP_004127.1 | 19 - 20 | 2631 | 114 | |
GenBank mRNA | AB209403.1 | - | 19 - 20 | 2564 | 114 |
AK027033.1 | - | 18 - 19 | 114 | ||
AK027034.1 | - | 10 - 11 | 1249 | 114 | |
AK292992.1 | - | 19 - 20 | 2515 | 114 | |
BC117481.1 | - | 19 - 20 | 2498 | 114 | |
BC117483.1 | - | 19 - 20 | 2498 | 114 | |
BC143948.1 | - | 18 - 19 | 2428 | 114 | |
M58511.1 | - | 19 - 20 | 2469 | 114 | |
U20180.1 | - | 18 - 19 | 2436 | 114 |