The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320941.1 | NP_001307870.1 | 20 - 21 | 2875 | 141 |
NM_001320942.1 | NP_001307871.1 | 21 - 22 | 3199 | 141 | |
NM_004136.3 | NP_004127.1 | 21 - 22 | 2945 | 141 | |
GenBank mRNA | AB209403.1 | - | 21 - 22 | 2878 | 141 |
AK027033.1 | - | 20 - 21 | 2709 | 141 | |
AK027034.1 | - | 12 - 13 | 1563 | 141 | |
AK292992.1 | - | 21 - 22 | 2829 | 141 | |
AL133439.1 | - | 2 - 3 | 246 | 141 | |
BC117481.1 | - | 21 - 22 | 2812 | 141 | |
BC117483.1 | - | 21 - 22 | 2812 | 141 | |
BC143948.1 | - | 20 - 21 | 2742 | 141 | |
BX340905.2 | - | 2 - 3 | 292 | 141 | |
M58511.1 | - | 21 - 22 | 2783 | 141 | |
U20180.1 | - | 20 - 21 | 2750 | 141 |