Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VWA7
Entrez Gene ID
80737
Gene Name
von Willebrand factor A domain containing 7
Gene Aliases
C6orf27, G7c, NG37
Chromosome Location
Chr.6: 31764135 - 31777331 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025258.2 NP_079534.2 9 - 10 1556 98
XM_005249427.2 XP_005249484.1 9 - 10 1556 98
XM_017011327.1 XP_016866816.1 8 - 9 1591 98
XM_017011328.1 XP_016866817.1 9 - 10 1702 98
XM_017011329.1 XP_016866818.1 7 - 8 1100 98
GenBank mRNA AJ245418.1 - 4 - 5 426 98
AK304058.1 - 9 - 10 1416 98
AK304266.1 - 8 - 9 1329 98
BC114625.1 - 8 - 9 1315 98

View Full Product Details