The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130182.1 | NP_001123654.1 | 3 - 4 | 541 | 81 |
NM_001130183.1 | NP_001123655.1 | 3 - 4 | 368 | 81 | |
NM_018602.3 | NP_061072.3 | 4 - 5 | 680 | 81 | |
XM_005254535.3 | XP_005254592.1 | 4 - 5 | 453 | 81 | |
XM_006720601.1 | XP_006720664.1 | 4 - 5 | 455 | 81 | |
GenBank mRNA | AF116663.1 | - | 2 - 3 | 772 | 81 |
AK096616.1 | - | 3 - 4 | 541 | 81 | |
AK098079.1 | - | 4 - 5 | 671 | 81 | |
BC021720.2 | - | 4 - 5 | 680 | 81 | |
BC031044.1 | - | 3 - 4 | 403 | 81 | |
BX648710.1 | - | 4 - 5 | 562 | 81 | |
DA561365.1 | - | 3 - 4 | 369 | 81 |