The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003253.2 | NP_003244.2 | 28 - 29 | 4778 | 87 |
XM_005261037.1 | XP_005261094.1 | 27 - 28 | 4911 | 87 | |
XM_005261038.2 | XP_005261095.1 | 26 - 27 | 4645 | 87 | |
XM_011529711.1 | XP_011528013.1 | 28 - 29 | 4862 | 87 | |
XM_011529712.1 | XP_011528014.1 | 28 - 29 | 4786 | 87 | |
XM_011529713.1 | XP_011528015.1 | 28 - 29 | 5196 | 87 | |
XM_017028447.1 | XP_016883936.1 | 28 - 29 | 4965 | 87 | |
XM_017028448.1 | XP_016883937.1 | 28 - 29 | 4935 | 87 | |
XM_017028449.1 | XP_016883938.1 | 27 - 28 | 4842 | 87 | |
XM_017028450.1 | XP_016883939.1 | 27 - 28 | 4862 | 87 | |
XM_017028451.1 | XP_016883940.1 | 27 - 28 | 5342 | 87 | |
XM_017028452.1 | XP_016883941.1 | 29 - 30 | 5142 | 87 | |
XM_017028453.1 | XP_016883942.1 | 27 - 28 | 4817 | 87 | |
GenBank mRNA | AB209101.1 | - | 24 - 25 | 3792 | 87 |
BC117192.1 | - | 25 - 26 | 4353 | 87 | |
BC117196.1 | - | 25 - 26 | 4353 | 87 | |
BC143980.1 | - | 23 - 24 | 4173 | 87 | |
U16296.1 | - | 28 - 29 | 4779 | 87 |