The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004337.2 | NP_004328.1 | 1 - 2 | 255 | 89 |
GenBank mRNA | AF061326.1 | - | 1 - 2 | 254 | 89 |