Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A32
Entrez Gene ID
81034
Gene Name
solute carrier family 25 member 32
Gene Aliases
MFT, MFTC, RREI
Chromosome Location
Chr.8: 103398638 - 103415335 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030780.4 NP_110407.2 6 - 7 1213 129
NR_102337.1 5 - 6 1127 129
NR_102338.1 7 - 8 1322 129
XM_017013877.1 XP_016869366.1 6 - 7 1116 129
GenBank mRNA AF283645.1 - 6 - 7 943 129
AK027531.1 - 6 - 7 982 129
AK027787.1 - 6 - 7 947 129
AK125492.1 - 7 - 8 931 129
AK300748.1 - 5 - 6 861 129
BC021893.1 - 6 - 7 921 129

View Full Product Details