The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006133.2 | NP_006124.1 | 4 - 5 | 530 | 61 |
XM_017018238.1 | XP_016873727.1 | 4 - 5 | 779 | 61 | |
XM_017018239.1 | XP_016873728.1 | 4 - 5 | 684 | 61 | |
XM_017018240.1 | XP_016873729.1 | 4 - 5 | 779 | 61 | |
GenBank mRNA | AB014559.2 | - | 4 - 5 | 525 | 61 |
AY275377.1 | - | 3 - 4 | 414 | 61 | |
BC150176.1 | - | 4 - 5 | 530 | 61 | |
BC150195.1 | - | 4 - 5 | 530 | 61 | |
BC152453.1 | - | 4 - 5 | 525 | 61 |