The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006133.2 | NP_006124.1 | 16 - 17 | 1770 | 81 |
XM_017018238.1 | XP_016873727.1 | 17 - 18 | 2097 | 81 | |
XM_017018239.1 | XP_016873728.1 | 17 - 18 | 2002 | 81 | |
GenBank mRNA | AB014559.2 | - | 16 - 17 | 1765 | 81 |
AY275377.1 | - | 15 - 16 | 1654 | 81 | |
BC150176.1 | - | 16 - 17 | 1770 | 81 | |
BC150195.1 | - | 16 - 17 | 1770 | 81 | |
BC152453.1 | - | 16 - 17 | 1765 | 81 |