Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C10orf67
Entrez Gene ID
256815
Gene Name
chromosome 10 open reading frame 67
Gene Aliases
C10orf115, LINC01552, bA215C7.4
Chromosome Location
Chr.10: 23202851 - 23345216 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011519440.1 XP_011517742.1 5 - 6 1138 155
XM_011519441.1 XP_011517743.1 5 - 6 1138 155
XM_011519442.2 XP_011517744.1 5 - 6 1138 155
XM_017016029.1 XP_016871518.1 5 - 6 1138 155
XM_017016030.1 XP_016871519.1 4 - 5 540 155
XM_017016031.1 XP_016871520.1 4 - 5 551 155

View Full Product Details