The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012197.3 | NP_036329.3 | 9 - 10 | 1337 | 153 |
XM_011518440.2 | XP_011516742.1 | 9 - 10 | 1341 | 153 | |
XM_011518441.2 | XP_011516743.1 | 10 - 11 | 1409 | 153 | |
XM_011518442.2 | XP_011516744.1 | 8 - 9 | 1164 | 153 | |
XM_011518444.2 | XP_011516746.1 | 9 - 10 | 1318 | 153 | |
XM_017014567.1 | XP_016870056.1 | 10 - 11 | 1626 | 153 | |
XM_017014568.1 | XP_016870057.1 | 10 - 11 | 1377 | 153 | |
XM_017014569.1 | XP_016870058.1 | 9 - 10 | 1439 | 153 | |
XM_017014570.1 | XP_016870059.1 | 8 - 9 | 1070 | 153 | |
GenBank mRNA | AB449897.1 | - | 8 - 9 | 1203 | 153 |
AJ011679.2 | - | 9 - 10 | 1308 | 153 | |
AK131449.1 | - | 10 - 11 | 1605 | 153 | |
BC020609.1 | - | 9 - 10 | 1337 | 153 | |
BC054492.1 | - | 9 - 10 | 1317 | 153 |