The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012197.3 | NP_036329.3 | 18 - 19 | 2428 | 94 |
XM_011518440.2 | XP_011516742.1 | 18 - 19 | 2432 | 94 | |
XM_011518441.2 | XP_011516743.1 | 19 - 20 | 2500 | 94 | |
XM_011518442.2 | XP_011516744.1 | 17 - 18 | 2255 | 94 | |
XM_017014567.1 | XP_016870056.1 | 19 - 20 | 2717 | 94 | |
XM_017014568.1 | XP_016870057.1 | 19 - 20 | 2468 | 94 | |
XM_017014569.1 | XP_016870058.1 | 18 - 19 | 2530 | 94 | |
XM_017014570.1 | XP_016870059.1 | 17 - 18 | 2161 | 94 | |
GenBank mRNA | AB449897.1 | - | 17 - 18 | 2294 | 94 |
AJ011679.2 | - | 18 - 19 | 2399 | 94 | |
AK131449.1 | - | 20 - 21 | 2926 | 94 | |
AK294531.1 | - | 6 - 7 | 734 | 94 | |
BC054492.1 | - | 18 - 19 | 2408 | 94 |