The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012197.3 | NP_036329.3 | 21 - 22 | 2764 | 76 |
XM_011518440.2 | XP_011516742.1 | 21 - 22 | 2768 | 76 | |
XM_011518441.2 | XP_011516743.1 | 22 - 23 | 2836 | 76 | |
XM_011518442.2 | XP_011516744.1 | 20 - 21 | 2591 | 76 | |
XM_017014567.1 | XP_016870056.1 | 22 - 23 | 3053 | 76 | |
XM_017014568.1 | XP_016870057.1 | 22 - 23 | 2804 | 76 | |
XM_017014569.1 | XP_016870058.1 | 21 - 22 | 2866 | 76 | |
XM_017014570.1 | XP_016870059.1 | 20 - 21 | 2497 | 76 | |
GenBank mRNA | AB449897.1 | - | 20 - 21 | 2630 | 76 |
AJ011679.2 | - | 21 - 22 | 2735 | 76 | |
AK131449.1 | - | 23 - 24 | 3262 | 76 | |
AK294531.1 | - | 9 - 10 | 1070 | 76 | |
AL050195.1 | - | 2 - 3 | 161 | 76 | |
BC054492.1 | - | 21 - 22 | 2744 | 76 |