The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012197.3 | NP_036329.3 | 24 - 25 | 3022 | 63 |
XM_011518440.2 | XP_011516742.1 | 24 - 25 | 3026 | 63 | |
XM_011518441.2 | XP_011516743.1 | 25 - 26 | 3094 | 63 | |
XM_011518442.2 | XP_011516744.1 | 23 - 24 | 2849 | 63 | |
XM_017014567.1 | XP_016870056.1 | 25 - 26 | 3311 | 63 | |
XM_017014568.1 | XP_016870057.1 | 25 - 26 | 3062 | 63 | |
XM_017014569.1 | XP_016870058.1 | 24 - 25 | 3124 | 63 | |
XM_017014570.1 | XP_016870059.1 | 23 - 24 | 2755 | 63 | |
GenBank mRNA | AB449897.1 | - | 23 - 24 | 2888 | 63 |
AJ011679.2 | - | 24 - 25 | 2993 | 63 | |
AK131449.1 | - | 26 - 27 | 3520 | 63 | |
AK294531.1 | - | 12 - 13 | 1328 | 63 | |
AL050195.1 | - | 5 - 6 | 419 | 63 | |
BC054492.1 | - | 24 - 25 | 3002 | 63 |