Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD29
Entrez Gene ID
147463
Gene Name
ankyrin repeat domain 29
Gene Aliases
-
Chromosome Location
Chr.18: 23598926 - 23662914 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308238.1 NP_001295167.1 8 - 9 877 99
XM_017025561.1 XP_016881050.1 7 - 8 800 99
GenBank mRNA AK057782.1 - 8 - 9 807 99

View Full Product Details