Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NMNAT3
Entrez Gene ID
349565
Gene Name
nicotinamide nucleotide adenylyltransferase 3
Gene Aliases
FKSG76, PNAT-3, PNAT3
Chromosome Location
Chr.3: 139560180 - 139678050 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178177.4 NP_835471.1 2 - 3 436 81
GenBank mRNA BC034374.1 - 2 - 3 384 81

View Full Product Details