The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099.4 | NP_001090.2 | 2 - 3 | 301 | 64 |
NM_001134194.1 | NP_001127666.1 | 2 - 3 | 301 | 64 | |
NM_001292037.1 | NP_001278966.1 | 2 - 3 | 301 | 64 | |
XM_011512946.1 | XP_011511248.1 | 2 - 3 | 265 | 64 | |
XM_011512947.2 | XP_011511249.1 | 2 - 3 | 265 | 64 | |
GenBank mRNA | AB102888.1 | - | 2 - 3 | 211 | 64 |
AK300540.1 | - | 2 - 3 | 262 | 64 | |
BC007460.1 | - | 2 - 3 | 261 | 64 | |
BC008493.1 | - | 2 - 3 | 258 | 64 | |
BC016344.1 | - | 2 - 3 | 265 | 64 | |
BC017877.1 | - | 2 - 3 | 251 | 64 | |
DC383453.1 | - | 2 - 3 | 301 | 64 | |
M24902.1 | - | 2 - 3 | 234 | 64 | |
M34840.1 | - | 2 - 3 | 225 | 64 |