The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002922.3 | NP_002913.3 | 2 - 3 | 284 | 88 |
GenBank mRNA | AF493925.1 | - | 2 - 3 | 179 | 88 |
AK093544.1 | - | 2 - 3 | 236 | 88 | |
AK303138.1 | - | 2 - 3 | 238 | 88 | |
AK315607.1 | - | 2 - 3 | 238 | 88 | |
AL039059.1 | - | 2 - 3 | 288 | 88 | |
BC015510.1 | - | 2 - 3 | 216 | 88 | |
BG536972.1 | - | 2 - 3 | 284 | 88 | |
BT006668.1 | - | 2 - 3 | 179 | 88 | |
EU668340.1 | - | 2 - 3 | 216 | 88 | |
X73427.1 | - | 2 - 3 | 193 | 88 |