Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CASP8AP2
Entrez Gene ID
9994
Gene Name
caspase 8 associated protein 2
Gene Aliases
CED-4, FLASH, RIP25
Chromosome Location
Chr.6: 89829900 - 89874436 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB037736.2 - - 6203 109
BC150274.1 - - 6232 109

View Full Product Details