The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015306.2 | NP_056121.2 | 50 - 51 | 6316 | 85 |
XM_005270690.3 | XP_005270747.1 | 37 - 38 | 4357 | 85 | |
XM_017000831.1 | XP_016856320.1 | 51 - 52 | 6381 | 85 | |
XM_017000832.1 | XP_016856321.1 | 51 - 52 | 6378 | 85 | |
XM_017000833.1 | XP_016856322.1 | 51 - 52 | 5873 | 85 | |
XM_017000834.1 | XP_016856323.1 | 50 - 51 | 5813 | 85 | |
GenBank mRNA | AB028980.1 | - | 9 - 10 | 1135 | 85 |
AK127075.1 | - | 33 - 34 | 4061 | 85 | |
AL832370.1 | - | 12 - 13 | 1542 | 85 |