Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DRD2
Entrez Gene ID
1813
Gene Name
dopamine receptor D2
Gene Aliases
D2DR, D2R
Chromosome Location
Chr.11: 113409595 - 113475279 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000795.3 NP_000786.1 1 - 2 201 94
NM_016574.3 NP_057658.2 1 - 2 201 94
GenBank mRNA BC021195.2 - 1 - 2 61 94
M30625.1 - 1 - 2 91
S69899.1 - 1 - 2 65 94
X51362.1 - 1 - 2 131 94

View Full Product Details