The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142640.1 | NP_001136112.1 | 6 - 7 | 3363 | 109 |
NM_018996.3 | NP_061869.2 | 6 - 7 | 3372 | 109 | |
XM_006721996.3 | XP_006722059.3 | 7 - 8 | 9024 | 109 | |
XM_006721997.3 | XP_006722060.3 | 7 - 8 | 9024 | 109 | |
XM_006722000.3 | XP_006722063.3 | 7 - 8 | 9015 | 109 | |
XM_011525088.2 | XP_011523390.1 | 4 - 5 | 3061 | 109 | |
XM_017024906.1 | XP_016880395.1 | 7 - 8 | 9015 | 109 | |
XM_017024907.1 | XP_016880396.1 | 7 - 8 | 9024 | 109 | |
XM_017024908.1 | XP_016880397.1 | 6 - 7 | 8799 | 109 | |
GenBank mRNA | AB046802.2 | - | 3 - 4 | 2922 | 109 |
AK098609.1 | - | 3 - 4 | 767 | 109 | |
AL834429.1 | - | 3 - 4 | 894 | 109 | |
BC045631.1 | - | 3 - 4 | 584 | 109 |