The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014010.4 | NP_054729.3 | 10 - 11 | 1989 | 68 |
GenBank mRNA | AB014534.1 | - | 10 - 11 | 1989 | 68 |
AF116574.1 | - | 8 - 9 | 1210 | 68 | |
AK127350.1 | - | 9 - 10 | 1210 | 68 | |
AK315025.1 | - | 8 - 9 | 1053 | 68 | |
BC018759.2 | - | 4 - 5 | 438 | 68 | |
BC146756.1 | - | 10 - 11 | 1989 | 68 | |
BX649176.1 | - | 10 - 11 | 1324 | 68 |