Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM266
Entrez Gene ID
123591
Gene Name
transmembrane protein 266
Gene Aliases
C15orf27, HVRP1
Chromosome Location
Chr.15: 76059929 - 76204963 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152335.2 NP_689548.2 4 - 5 531 84
XM_017021915.1 XP_016877404.1 6 - 7 747 84
GenBank mRNA BX398159.2 - 3 - 4 447 84
DA090797.1 - 4 - 5 504 84

View Full Product Details