The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160011.1 | NP_001153483.1 | 3 - 4 | 789 | 94 |
XM_017000939.1 | XP_016856428.1 | 3 - 4 | 3941 | 94 | |
GenBank mRNA | AY358540.1 | - | 3 - 4 | 748 | 94 |
BC039520.1 | - | 2 - 3 | 500 | 94 |