Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZMYND15
Entrez Gene ID
84225
Gene Name
zinc finger MYND-type containing 15
Gene Aliases
SPGF14
Chromosome Location
Chr.17: 4740015 - 4746119 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136046.2 NP_001129518.1 13 - 14 58
NM_001267822.1 NP_001254751.1 13 - 14 58
NM_032265.2 NP_115641.1 12 - 13 58
XM_017025218.1 XP_016880707.1 10 - 11 58
GenBank mRNA AA937353.1 - 1 - 2 299 58
AK302181.1 - 13 - 14 58
AL136893.1 - 11 - 12 58

View Full Product Details