The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021937.4 | NP_068756.2 | 6 - 7 | 1649 | 68 |
XM_005247696.2 | XP_005247753.1 | 5 - 6 | 1485 | 68 | |
XM_011513066.2 | XP_011511368.1 | 4 - 5 | 1097 | 68 | |
XM_017006978.1 | XP_016862467.1 | 7 - 8 | 1429 | 68 | |
XM_017006979.1 | XP_016862468.1 | 6 - 7 | 1918 | 68 | |
XM_017006981.1 | XP_016862470.1 | 3 - 4 | 1444 | 68 | |
GenBank mRNA | AF268872.1 | - | 6 - 7 | 1388 | 68 |
BC007933.2 | - | 6 - 7 | 1527 | 68 |