Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPO
Entrez Gene ID
130749
Gene Name
carboxypeptidase O
Gene Aliases
-
Chromosome Location
Chr.2: 206932853 - 206971064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173077.2 NP_775100.1 1 - 2 112 107
XM_017003372.1 XP_016858861.1 2 - 3 6219 107
GenBank mRNA AJ422118.1 - 1 - 2 66 107
BC112076.1 - 1 - 2 112 107
BC112078.1 - 1 - 2 112 107

View Full Product Details