Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRIPAP1
Entrez Gene ID
56850
Gene Name
GRIP1 associated protein 1
Gene Aliases
GRASP-1
Chromosome Location
Chr.X: 48973720 - 49002264 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020137.4 NP_064522.3 14 - 15 1207 61
XM_011543935.1 XP_011542237.1 14 - 15 1207 61
XM_011543936.1 XP_011542238.1 14 - 15 1207 61
XM_011543937.1 XP_011542239.1 14 - 15 1254 61
XM_017029656.1 XP_016885145.1 14 - 15 1207 61
XM_017029657.1 XP_016885146.1 14 - 15 1578 61
XM_017029658.1 XP_016885147.1 14 - 15 2517 61
XM_017029659.1 XP_016885148.1 14 - 15 1254 61
XM_017029660.1 XP_016885149.1 14 - 15 1608 61
XM_017029661.1 XP_016885150.1 14 - 15 1254 61
GenBank mRNA AB032993.1 - 14 - 15 1152 61
AK292158.1 - 14 - 15 1178 61
AL136847.1 - 1 - 2 93 61
BC001522.2 - 4 - 5 320 61
BC101544.1 - 14 - 15 1205 61
BC101546.1 - 14 - 15 1205 61
BX640704.1 - 8 - 9 891 61
BX647801.1 - 13 - 14 1047 61
BX647804.1 - 13 - 14 1047 61

View Full Product Details