The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020137.4 | NP_064522.3 | 8 - 9 | 727 | 118 |
XM_011543935.1 | XP_011542237.1 | 8 - 9 | 727 | 118 | |
XM_011543936.1 | XP_011542238.1 | 8 - 9 | 727 | 118 | |
XM_011543937.1 | XP_011542239.1 | 8 - 9 | 774 | 118 | |
XM_017029656.1 | XP_016885145.1 | 8 - 9 | 727 | 118 | |
XM_017029657.1 | XP_016885146.1 | 8 - 9 | 1098 | 118 | |
XM_017029658.1 | XP_016885147.1 | 8 - 9 | 2037 | 118 | |
XM_017029659.1 | XP_016885148.1 | 8 - 9 | 774 | 118 | |
XM_017029660.1 | XP_016885149.1 | 8 - 9 | 1128 | 118 | |
XM_017029661.1 | XP_016885150.1 | 8 - 9 | 774 | 118 | |
GenBank mRNA | AB032993.1 | - | 8 - 9 | 672 | 118 |
AK292158.1 | - | 8 - 9 | 698 | 118 | |
BC101544.1 | - | 8 - 9 | 725 | 118 | |
BC101546.1 | - | 8 - 9 | 725 | 118 | |
BX640704.1 | - | 2 - 3 | 411 | 118 | |
BX647801.1 | - | 7 - 8 | 567 | 118 | |
BX647804.1 | - | 7 - 8 | 567 | 118 |