The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032492.3 | NP_115881.3 | 1 - 2 | 214 | 110 |
GenBank mRNA | AK027508.1 | - | 1 - 2 | 160 | 110 |
AK074760.1 | - | 1 - 2 | 213 | 110 | |
AK225756.1 | - | 1 - 2 | 196 | 110 | |
BC032101.2 | - | 1 - 2 | 194 | 110 | |
BP262753.1 | - | 1 - 2 | 229 | 110 |