Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C2orf47
Entrez Gene ID
79568
Gene Name
chromosome 2 open reading frame 47
Gene Aliases
-
Chromosome Location
Chr.2: 199955317 - 199964125 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024520.2 NP_078796.2 3 - 4 848 131
XM_005246852.2 XP_005246909.1 2 - 3 759 131
GenBank mRNA AK026208.1 - 3 - 4 848 131
AL832964.1 - 2 - 3 167 131
BC017959.1 - 2 - 3 532 131

View Full Product Details