The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040428.3 | NP_001035518.1 | 5 - 6 | 914 | 127 |
NM_018418.4 | NP_060888.2 | 6 - 7 | 1010 | 127 | |
XM_005267851.1 | XP_005267908.1 | 6 - 7 | 981 | 127 | |
XM_005267852.1 | XP_005267909.1 | 5 - 6 | 910 | 127 | |
XM_005267854.1 | XP_005267911.1 | 7 - 8 | 1043 | 127 | |
XM_005267855.1 | XP_005267912.1 | 6 - 7 | 976 | 127 | |
XM_006720204.1 | XP_006720267.1 | 6 - 7 | 981 | 127 | |
XM_006720205.1 | XP_006720268.1 | 6 - 7 | 981 | 127 | |
XM_011536951.1 | XP_011535253.1 | 6 - 7 | 1175 | 127 | |
XM_011536952.1 | XP_011535254.1 | 4 - 5 | 793 | 127 | |
XM_011536953.1 | XP_011535255.1 | 5 - 6 | 1114 | 127 | |
XM_017021452.1 | XP_016876941.1 | 6 - 7 | 1175 | 127 | |
XM_017021453.1 | XP_016876942.1 | 6 - 7 | 1141 | 127 | |
XM_017021454.1 | XP_016876943.1 | 7 - 8 | 1043 | 127 | |
XM_017021455.1 | XP_016876944.1 | 6 - 7 | 976 | 127 | |
XM_017021456.1 | XP_016876945.1 | 7 - 8 | 989 | 127 | |
XM_017021457.1 | XP_016876946.1 | 5 - 6 | 1114 | 127 | |
GenBank mRNA | AF144487.3 | - | 6 - 7 | 990 | 127 |
AF144488.2 | - | 4 - 5 | 715 | 127 | |
AK055864.1 | - | 6 - 7 | 932 | 127 | |
AK225746.1 | - | 7 - 8 | 981 | 127 | |
AK290631.1 | - | 5 - 6 | 885 | 127 | |
AK293007.1 | - | 7 - 8 | 963 | 127 | |
AL136604.1 | - | 5 - 6 | 894 | 127 | |
BC008656.1 | - | 1 - 2 | 442 | 127 | |
BC090875.1 | - | 6 - 7 | 1297 | 127 | |
CR533563.1 | - | 5 - 6 | 739 | 127 | |
CR936777.1 | - | 6 - 7 | 956 | 127 | |
EU668353.1 | - | 6 - 7 | 986 | 127 |