The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004522.2 | NP_004513.1 | 7 - 8 | 988 | 138 |
XM_017004062.1 | XP_016859551.1 | 7 - 8 | 979 | 138 | |
GenBank mRNA | AB011103.1 | - | 7 - 8 | 961 | 138 |
BC002721.2 | - | 7 - 8 | 970 | 138 | |
BC017298.2 | - | 7 - 8 | 943 | 138 | |
BC025961.1 | - | 7 - 8 | 970 | 138 | |
BC127772.1 | - | 4 - 5 | 312 | 138 |