Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG4A
Entrez Gene ID
115201
Gene Name
autophagy related 4A cysteine peptidase
Gene Aliases
APG4A, AUTL2
Chromosome Location
Chr.X: 108088900 - 108154671 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321287.1 NP_001308216.1 6 - 7 1276 123
NM_001321288.1 NP_001308217.1 5 - 6 597 123
NM_001321289.1 NP_001308218.1 6 - 7 856 123
NM_001321290.1 NP_001308219.1 6 - 7 856 123
NM_052936.4 NP_443168.2 5 - 6 554 123
NM_178270.3 NP_840054.1 5 - 6 554 123
NM_178271.2 NP_840055.1 6 - 7 856 123
NR_135608.1 5 - 6 554 123
XM_011530841.2 XP_011529143.1 5 - 6 455 123
XM_011530842.1 XP_011529144.1 6 - 7 782 123
GenBank mRNA AB066214.1 - 4 - 5 394 123
AJ320508.1 - 5 - 6 412 123
AJ320509.1 - 5 - 6 412 123
AJ504651.1 - 5 - 6 423 123
AK293533.1 - 6 - 7 735 123
AK314429.1 - 5 - 6 421 123
BC041862.1 - 6 - 7 739 123
BC061696.1 - 5 - 6 556 123

View Full Product Details