The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015529.3 | NP_056344.2 | 8 - 9 | 1416 | 149 |
XM_017010714.1 | XP_016866203.1 | 8 - 9 | 1224 | 149 | |
XM_017010715.1 | XP_016866204.1 | 8 - 9 | 1062 | 149 | |
GenBank mRNA | AK074879.1 | - | 8 - 9 | 1131 | 149 |
AK226054.1 | - | 4 - 5 | 513 | 149 | |
AY007239.1 | - | 7 - 8 | 1332 | 149 | |
AY359094.1 | - | 8 - 9 | 1340 | 149 | |
BC018756.1 | - | 8 - 9 | 1375 | 149 |