The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079521.1 | NP_001072989.1 | 15 - 16 | 2162 | 70 |
NM_001079522.1 | NP_001072990.1 | 16 - 17 | 2286 | 70 | |
NM_001271014.1 | NP_001257943.1 | 16 - 17 | 2286 | 70 | |
NM_004986.3 | NP_004977.2 | 15 - 16 | 2162 | 70 | |
NR_073128.1 | 16 - 17 | 2286 | 70 | ||
NR_073129.1 | 16 - 17 | 2286 | 70 | ||
XM_006720138.2 | XP_006720201.1 | 16 - 17 | 2277 | 70 | |
XM_006720139.2 | XP_006720202.1 | 15 - 16 | 2125 | 70 | |
XM_006720140.3 | XP_006720203.1 | 15 - 16 | 2125 | 70 | |
XM_006720141.2 | XP_006720204.1 | 15 - 16 | 2125 | 70 | |
XM_011536751.2 | XP_011535053.1 | 15 - 16 | 2383 | 70 | |
XM_011536752.1 | XP_011535054.1 | 15 - 16 | 2125 | 70 | |
XM_011536753.1 | XP_011535055.1 | 15 - 16 | 2125 | 70 | |
XM_017021278.1 | XP_016876767.1 | 15 - 16 | 7975 | 70 | |
XM_017021279.1 | XP_016876768.1 | 15 - 16 | 2125 | 70 | |
XM_017021280.1 | XP_016876769.1 | 16 - 17 | 2277 | 70 | |
XM_017021281.1 | XP_016876770.1 | 16 - 17 | 8099 | 70 | |
XM_017021282.1 | XP_016876771.1 | 16 - 17 | 2280 | 70 | |
XM_017021283.1 | XP_016876772.1 | 16 - 17 | 2214 | 70 | |
XM_017021284.1 | XP_016876773.1 | 15 - 16 | 2125 | 70 | |
XM_017021285.1 | XP_016876774.1 | 16 - 17 | 2257 | 70 | |
XM_017021286.1 | XP_016876775.1 | 15 - 16 | 2125 | 70 | |
GenBank mRNA | AK302797.1 | - | 16 - 17 | 2240 | 70 |
AY264265.1 | - | 16 - 17 | 2171 | 70 | |
AY536375.1 | - | 5 - 6 | 375 | 70 | |
BC050555.1 | - | 15 - 16 | 2101 | 70 | |
BC112337.1 | - | 16 - 17 | 2169 | 70 | |
BC117132.1 | - | 15 - 16 | 2103 | 70 | |
BC143720.1 | - | 15 - 16 | 2103 | 70 | |
D13629.1 | - | 15 - 16 | 2067 | 70 | |
L25616.1 | - | 14 - 15 | 2024 | 70 | |
Z22551.1 | - | 15 - 16 | 2050 | 70 |