Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IZUMO4
Entrez Gene ID
113177
Gene Name
IZUMO family member 4
Gene Aliases
C19orf36, IMAGE:4215339
Chromosome Location
Chr.19: 2096848 - 2099590 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031735.2 NP_001026905.2 6 - 7 591 74
NM_001039846.1 NP_001034935.1 6 - 7 591 74
GenBank mRNA AY753639.1 - 6 - 7 550 74
BC014609.2 - 6 - 7 545 74
BI827974.1 - 6 - 7 73
BX378854.2 - 6 - 7 591 74
DQ196309.1 - 6 - 7 578 74
DQ196310.1 - 6 - 7 578 74
DQ424900.1 - 6 - 7 514 74

View Full Product Details