The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085487.2 | NP_001078956.1 | 9 - 10 | 1411 | 123 |
XM_006710314.3 | XP_006710377.1 | 9 - 10 | 1427 | 123 | |
XM_011540573.2 | XP_011538875.1 | 10 - 11 | 1391 | 123 | |
XM_011540574.2 | XP_011538876.1 | 9 - 10 | 1659 | 123 | |
XM_017000195.1 | XP_016855684.1 | 10 - 11 | 1463 | 123 | |
XM_017000196.1 | XP_016855685.1 | 7 - 8 | 1313 | 123 | |
XM_017000197.1 | XP_016855686.1 | 5 - 6 | 1091 | 123 | |
GenBank mRNA | AB067502.1 | - | 5 - 6 | 1437 | 123 |
AK126835.1 | - | 3 - 4 | 960 | 123 | |
BX648168.1 | - | 6 - 7 | 1147 | 123 |