The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005468.2 | NP_005459.2 | 10 - 11 | 1365 | 59 |
XM_011544706.2 | XP_011543008.1 | 10 - 11 | 1685 | 59 | |
XM_011544707.2 | XP_011543009.1 | 11 - 12 | 1333 | 59 | |
XM_011544709.2 | XP_011543011.1 | 10 - 11 | 1705 | 59 | |
XM_011544710.2 | XP_011543012.1 | 10 - 11 | 1702 | 59 | |
GenBank mRNA | AF010141.1 | - | 1 - 2 | 95 | 59 |
AJ012371.1 | - | 10 - 11 | 1365 | 59 |