The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005468.2 | NP_005459.2 | 16 - 17 | 1960 | 81 |
XM_011544706.2 | XP_011543008.1 | 14 - 15 | 2271 | 81 | |
XM_011544707.2 | XP_011543009.1 | 17 - 18 | 1928 | 81 | |
XM_011544708.2 | XP_011543010.1 | 14 - 15 | 2121 | 81 | |
GenBank mRNA | AF010141.1 | - | 7 - 8 | 690 | 81 |
AJ012371.1 | - | 16 - 17 | 1960 | 81 | |
AK311090.1 | - | 5 - 6 | 577 | 81 | |
BG757994.1 | - | 4 - 5 | 436 | 81 |