Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAALADL1
Entrez Gene ID
10004
Gene Name
N-acetylated alpha-linked acidic dipeptidase-like 1
Gene Aliases
I100, NAALADASEL
Chromosome Location
Chr.11: 65044818 - 65059020 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005468.2 NP_005459.2 7 - 8 1088 65
XM_011544706.2 XP_011543008.1 7 - 8 1408 65
XM_011544707.2 XP_011543009.1 8 - 9 1056 65
XM_011544708.2 XP_011543010.1 7 - 8 1408 65
XM_011544709.2 XP_011543011.1 7 - 8 1428 65
XM_011544710.2 XP_011543012.1 7 - 8 1425 65
XM_011544711.2 XP_011543013.1 7 - 8 1430 65
XM_011544712.2 XP_011543014.1 7 - 8 1431 65
GenBank mRNA AJ012371.1 - 7 - 8 1088 65

View Full Product Details