Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FFAR1
- Entrez Gene ID
-
2864
- Gene Name
- free fatty acid receptor 1
- Gene Aliases
-
FFA1R, GPCR40, GPR40
- Chromosome Location
-
Chr.19: 35351542 - 35352464 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC039436.1 |
- |
1 - 1 |
399 |
67 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BI603605.1 |
- |
1 |
404 |
|
Chr. 19 - 35352419 - 35353229 [+] on Build GRCh38 |
67 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
CD22 molecule |
CD22 |
SIGLEC-2, SIGLEC2 |
-5.1kb
|
Sense |
Chr. - 35329166 - 35347361 on Build GRCh38 |
Downstream |
free fatty acid receptor 3 |
FFAR3 |
FFA3R, GPR41, GPR42 |
5.0kb
|
Sense |
Chr. - 35358106 - 35360491 on Build GRCh38 |
View Full Product Details