The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307930.1 | NP_001294859.1 | 2 - 3 | 453 | 56 |
NM_016563.3 | NP_057647.1 | 3 - 4 | 510 | 56 | |
XM_005254434.4 | XP_005254491.1 | 3 - 4 | 320 | 56 | |
XM_011521660.2 | XP_011519962.1 | 4 - 5 | 323 | 56 | |
XM_017022296.1 | XP_016877785.1 | 3 - 4 | 320 | 56 | |
GenBank mRNA | AF233588.1 | - | 3 - 4 | 349 | 56 |
AK296261.1 | - | 2 - 3 | 263 | 56 | |
AK300536.1 | - | 3 - 4 | 293 | 56 | |
AK314914.1 | - | 3 - 4 | 318 | 56 | |
BC053734.1 | - | 3 - 4 | 358 | 56 |