The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012335.3 | NP_036467.2 | 7 - 8 | 773 | 93 |
XM_011528024.1 | XP_011526326.1 | 7 - 8 | 773 | 93 | |
XM_011528025.1 | XP_011526327.1 | 7 - 8 | 773 | 93 | |
XM_011528026.1 | XP_011526328.1 | 7 - 8 | 773 | 93 | |
XM_011528027.1 | XP_011526329.1 | 7 - 8 | 773 | 93 | |
XM_011528028.2 | XP_011526330.1 | 7 - 8 | 773 | 93 | |
XM_017026821.1 | XP_016882310.1 | 7 - 8 | 773 | 93 | |
GenBank mRNA | AB210037.1 | - | 7 - 8 | 772 | 93 |
AB290178.1 | - | 7 - 8 | 903 | 93 | |
AJ310570.1 | - | 7 - 8 | 675 | 93 | |
AK131106.1 | - | 7 - 8 | 772 | 93 | |
BC028071.1 | - | 7 - 8 | 749 | 93 |