Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADCK5
Entrez Gene ID
203054
Gene Name
aarF domain containing kinase 5
Gene Aliases
-
Chromosome Location
Chr.8: 144373762 - 144393242 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174922.4 NP_777582.4 2 - 3 195 144
XM_006716527.2 XP_006716590.1 2 - 3 190 144
XM_011516907.2 XP_011515209.1 2 - 3 908 144
XM_011516909.1 XP_011515211.1 2 - 3 195 144
XM_011516910.1 XP_011515212.1 2 - 3 149 144
XM_017013174.1 XP_016868663.1 2 - 3 233 144
GenBank mRNA AK092773.1 - 2 - 3 158 144
BC101659.1 - 2 - 3 135 144
BC143326.1 - 2 - 3 135 144
BM146288.1 - 2 - 3 181 144
HY037845.1 - 3 - 4 293 144

View Full Product Details