Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- C11orf49
- Entrez Gene ID
-
79096
- Gene Name
- chromosome 11 open reading frame 49
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 46936524 - 47164381 on Build GRCh38
View Full Product Details