The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018126.2 | NP_060596.2 | 5 - 6 | 901 | 119 |
XM_005248116.3 | XP_005248173.1 | 6 - 7 | 740 | 119 | |
XM_005248117.2 | XP_005248174.1 | 6 - 7 | 688 | 119 | |
GenBank mRNA | AK001387.1 | - | 5 - 6 | 869 | 119 |
AK023062.1 | - | 5 - 6 | 881 | 119 | |
AK094190.1 | - | 6 - 7 | 812 | 119 | |
AK308501.1 | - | 4 - 5 | 681 | 119 | |
AY659966.1 | - | 5 - 6 | 869 | 119 | |
BC000948.2 | - | 5 - 6 | 631 | 119 | |
CR457260.1 | - | 5 - 6 | 536 | 119 |